1-Brom-4-(difluormethoxy)benzol
Lieferant: COMBI-BLOCKS
Synonyme:
4-(Difluoromethoxy)bromobenzene
OS-7439-100G
OS-7439-5G
OS-7439-25G
COBBOS-7439-100GEA
374
EUR
COBBOS-7439-100G
COBBOS-7439-5G
COBBOS-7439-25G
1-Brom-4-(difluormethoxy)benzol
1-Brom-4-(difluormethoxy)benzol
Formel:
C₇H₅BrF₂O Molecular Weight: 223.02 g/mol |
MDL:
MFCD00221465 CAS-Nummer: 5905-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...