4-Brombenzylalkohol

Lieferant: COMBI-BLOCKS

Synonyme: (4-Bromophenyl)methanol

OS-7636-100G OS-7636-5G
COBBOS-7636-100GEA 141 EUR
COBBOS-7636-100G COBBOS-7636-5G
4-Brombenzylalkohol
4-Brombenzylalkohol
Formel: C₇H₇BrO
Molecular Weight: 187,04 g/mol
Siedepunkt: 268 °C (1013 hPa)
Schmelzpunkt: 77…78 °C
MDL: MFCD00004650
CAS-Nummer: 873-75-6
EINECS: 212-851-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR