4-Bromcumol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromoisopropylbenzene, 1-Bromo-4-isopropylbenzene

OT-0600-5G OT-0600-25G OT-0600-100G
COBBOT-0600-5GEA 48.4 EUR
COBBOT-0600-5G COBBOT-0600-25G COBBOT-0600-100G
4-Bromcumol
4-Bromcumol
Formel: C₉H₁₁Br
Molecular Weight: 199,09 g/mol
MDL: MFCD00039159
CAS-Nummer: 586-61-8
EINECS: 209-577-5
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR