1-Brom-4-chlor-2-nitrobenzol

Lieferant: COMBI-BLOCKS
Achtung

AN-1055-25G AN-1055-5G AN-1055-100G
COBBAN-1055-25GEA 51.3 EUR
COBBAN-1055-25G COBBAN-1055-5G COBBAN-1055-100G
1-Brom-4-chlor-2-nitrobenzol
1-Brom-4-chlor-2-nitrobenzol
Formel: C₆H₃BrClNO₂
Molecular Weight: 236,45 g/mol
Schmelzpunkt: 70 °C
Dichte: 2,048 g/cm³ (20 °C)
MDL: MFCD00024320
CAS-Nummer: 41513-04-6
EINECS: 255-421-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR