1-Brom-4-chlor-3-fluorbenzol

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 4-Bromo-1-chloro-2-fluorobenzene

OS-7699-5G OS-7699-25G OS-7699-100G
COBBOS-7699-5GEA 25.8 EUR
COBBOS-7699-5G COBBOS-7699-25G COBBOS-7699-100G
1-Brom-4-chlor-3-fluorbenzol
1-Brom-4-chlor-3-fluorbenzol
Formel: C₆H₃BrClF
Molecular Weight: 209.44 g/mol
Siedepunkt: 130…142 °C (20 mmHg)
Dichte: 1.71 g/cm³ (20 °C)
MDL: MFCD00042519
CAS-Nummer: 60811-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR