4-Brom-3-methoxybenzonitril

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-3-methoxybenzoesäurenitril

CA-4369-1G CA-4369-5G CA-4369-25G
COBBCA-4369-1GEA 38.2 EUR
COBBCA-4369-1G COBBCA-4369-5G COBBCA-4369-25G
4-Brom-3-methoxybenzonitril
4-Brom-3-methoxybenzonitril
Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
MDL: MFCD09753723
CAS-Nummer: 120315-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR