4-Brom-2-fluorbenzonitril 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Brom-4-cyan-3-fluorbenzene
290490050
290490010
290490250
ACRO290490050EA
58.8
EUR
ACRO290490050
ACRO290490010
ACRO290490250
4-Brom-2-fluorbenzonitril 99+%
4-Brom-2-fluorbenzonitril
Formel:
C₇H₃BrFN Molecular Weight: 200.01 g/mol Siedepunkt: 238…239 °C (1013 hPa) Schmelzpunkt: 69…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143264 CAS-Nummer: 105942-08-3 UN: 3439 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | White to light brown Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 69 to 72 °C |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...