4-Brom-2-fluorbenzonitril

Lieferant: COMBI-BLOCKS

Synonyme: 1-Brom-4-cyan-3-fluorbenzene

OS-7559-25G OS-7559-100G
COBBOS-7559-25GEA 70 EUR
COBBOS-7559-25G COBBOS-7559-100G
4-Brom-2-fluorbenzonitril
4-Brom-2-fluorbenzonitril
Formel: C₇H₃BrFN
Molecular Weight: 200.01 g/mol
Siedepunkt: 238…239 °C (1013 hPa)
Schmelzpunkt: 71…73 °C
MDL: MFCD00143264
CAS-Nummer: 105942-08-3
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR