4-Brom-2-methoxybenzonitril
Lieferant: COMBI-BLOCKS
Synonyme:
4-Brom-2-methoxybenzoesäurenitril
CA-4677-5G
CA-4677-1G
COBBCA-4677-5GEA
75.9
EUR
COBBCA-4677-5G
COBBCA-4677-1G
4-Brom-2-methoxybenzonitril
4-Brom-2-methoxybenzonitril
Formel:
C₈H₆BrNO Molecular Weight: 212,05 g/mol |
MDL:
MFCD06738651 CAS-Nummer: 330793-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...