4-Brom-o-toluonitril

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-2-methylbenzonitrile

OS-7570-5G OS-7570-25G
COBBOS-7570-5GEA 50.9 EUR
COBBOS-7570-5G COBBOS-7570-25G
4-Brom-o-toluonitril
4-Brom-o-toluonitril
Formel: C₈H₆BrN
Molecular Weight: 196,05 g/mol
Siedepunkt: 276 °C (1013 hPa)
Schmelzpunkt: 70 °C
MDL: MFCD00041461
CAS-Nummer: 67832-11-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR