5-Brom-2-fluorphenol

Lieferant: COMBI-BLOCKS

OT-0190-5G OT-0190-25G OT-0190-100G
COBBOT-0190-5GEA 44.5 EUR
COBBOT-0190-5G COBBOT-0190-25G COBBOT-0190-100G
5-Brom-2-fluorphenol
5-Brom-2-fluorphenol
Formel: C₆H₄BrFO
Molecular Weight: 191 g/mol
MDL: MFCD07782066
CAS-Nummer: 112204-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR