1-Brom-4-fluor-3-nitrobenzol
Lieferant: COMBI-BLOCKS
Synonyme:
5-Brom-2-fluornitrobenzol, 4-Brom-1-fluor-2-nitrobenzol
OS-7448-5G
OS-7448-100G
OS-7448-25G
COBBOS-7448-5GEA
0
EUR
COBBOS-7448-5G
COBBOS-7448-100G
COBBOS-7448-25G
1-Brom-4-fluor-3-nitrobenzol
1-Bromo-4-fluoro-3-nitrobenzene
Formel:
C₆H₃BrFNO₂ Molecular Weight: 220 g/mol Siedepunkt: 145…147 °C (35 mmHg) Schmelzpunkt: 19 Dichte: 1.786 g/cm³ (20 °C) |
MDL:
MFCD00129165 CAS-Nummer: 364-73-8 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...