5-Brom-2-hydroxybenzylalkohol
Lieferant: Thermo Scientific
Synonyme:
4-Brom-2-(hydroxymethyl)phenol, (5-Brom-2-hydroxyphenyl)methanol
CD03713.1
CD03713.10
CD03713.50
MAYBCD03713.1EA
0
EUR
MAYBCD03713.1
MAYBCD03713.10
MAYBCD03713.50
5-Brom-2-hydroxybenzylalkohol
5-Brom-2-hydroxybenzylalkohol
Formel:
C₇H₇BrO₂ Siedepunkt: 302 °C (1013 hPa) Schmelzpunkt: 113 °C |
MDL:
MFCD00004618 CAS-Nummer: 2316-64-5 EINECS: 219-026-0 Merck Index: 12,01460 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...