4-Brom-2-(trifluormethyl)phenol
Lieferant: BIONET RESEARCH
Synonyme:
5-Bromo-2-hydroxybenzotrifluoride
9X-0882
9X-0882.1
BIOT9X-0882EA
0
EUR
BIOT9X-0882
BIOT9X-0882.1
4-Brom-2-(trifluormethyl)phenol
4-Brom-2-(trifluormethyl)phenol
Formel:
C₇H₄BrF₃O |
MDL:
MFCD05668981 CAS-Nummer: 50824-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...