4-Brom-2-methoxyphenol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromguajacol, 1-Brom-4-hydroxy-3-methoxybenzol, 5-Brom-2-hydroxyanisol

OT-0428-1G OT-0428-25G OT-0428-5G
COBBOT-0428-1GEA 22 EUR
COBBOT-0428-1G COBBOT-0428-25G COBBOT-0428-5G
4-Brom-2-methoxyphenol
4-Brom-2-methoxyphenol
Formel: C₇H₇BrO₂
Molecular Weight: 203.04 g/mol
MDL: MFCD00051937
CAS-Nummer: 7368-78-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR