5-Brom-2-iodbenzotrifluorid
Lieferant: COMBI-BLOCKS
Synonyme:
4-Brom-2-(trifluormethyl)iodbenzol, 5-Brom-2-iodbenzotrifluorid, 1-Brom-4-iod-3-(trifluormethyl)benzol
HC-2384-5G
HC-2384-25G
HC-2384-1G
HC-2384-100G
COBBHC-2384-5GEA
33.2
EUR
COBBHC-2384-5G
COBBHC-2384-25G
COBBHC-2384-1G
COBBHC-2384-100G
5-Brom-2-iodbenzotrifluorid
5-Brom-2-iodbenzotrifluorid
Formel:
C₇H₃BrF₃I Molecular Weight: 350.9 |
MDL:
MFCD00672935 CAS-Nummer: 364-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...