1-Brom-4-iodbenzol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
4-Bromo-1-iodobenzene
OS-7533-25G
OS-7533-100G
COBBOS-7533-25GEA
50.3
EUR
COBBOS-7533-25G
COBBOS-7533-100G
1-Brom-4-iodbenzol
1-Brom-4-iodbenzol
Formel:
C₆H₄BrI Molecular Weight: 282,91 g/mol Siedepunkt: 245 °C (1013 hPa) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001051 CAS-Nummer: 589-87-7 EINECS: 209-662-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...