4-Brom-3-methylanisol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-5-methoxytoluol, 1-Brom-4-methoxy-2-methylbenzol

OT-0842-1G OT-0842-25G OT-0842-5G
COBBOT-0842-1GEA 0 EUR
COBBOT-0842-1G COBBOT-0842-25G COBBOT-0842-5G
4-Brom-3-methylanisol
4-Brom-3-methylanisol
Formel: C₈H₉BrO
Molecular Weight: 201.06 g/mol
MDL: MFCD00060687
CAS-Nummer: 27060-75-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR