4-Brom-3-nitroanisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-4-methoxy-2-nitrobenzene

OT-0054-5G OT-0054-25G OT-0054-100G
COBBOT-0054-5GEA 0 EUR
COBBOT-0054-5G COBBOT-0054-25G COBBOT-0054-100G
4-Brom-3-nitroanisol
4-Brom-3-nitroanisol
Formel: C₇H₆BrNO₃
Molecular Weight: 232.03 g/mol
MDL: MFCD00051511
CAS-Nummer: 5344-78-5
EINECS: 226-290-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR