1-Brom-4-pentylbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-pentylbenzol, (4-Bromphenyl)pentan, p-Brom-pentylbenzol

OS-7594-5G OS-7594-25G OS-7594-100G
COBBOS-7594-5GEA 26.2 EUR
COBBOS-7594-5G COBBOS-7594-25G COBBOS-7594-100G
1-Brom-4-pentylbenzol
1-Brom-4-pentylbenzol
Formel: C₁₁H₁₅Br
Molecular Weight: 227,14 g/mol
MDL: MFCD00061113
CAS-Nummer: 51554-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR