3'-Chlor-2',6'-difluoracetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(3-Chloro-2,6-difluorophenyl)ethanone
005583-1G
005583-5G
FLUO005583-1GEA
57.9
EUR
FLUO005583-1G
FLUO005583-5G
3'-Chlor-2',6'-difluoracetophenon
3'-Chlor-2',6'-difluoracetophenon
Formel:
C₈H₅ClF₂O Molecular Weight: 190.58 g/mol |
MDL:
MFCD01631329 CAS-Nummer: 177942-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...