4'-Chlor-2',5'-difluoracetophenon

Lieferant: CHEMPUR

FL1626.1 001626-5G 001626-25G
CHMPFL1626.1EA 0 EUR
CHMPFL1626.1 FLUO001626-5G FLUO001626-25G
4'-Chlor-2',5'-difluoracetophenon
4'-Chlor-2',5'-difluoracetophenon
Formel: C₈H₅ClF₂O
MDL: MFCD00068247
CAS-Nummer: 655-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR