1-Chlor-2,6-difluor-3-nitrobenzol

Lieferant: CHEMPUR

Synonyme: 2-Chlor-1,3-difluor-4-nitrobenzol, 3-Chlor-2,4-difluornitrobenzol

JFD00909.250 JFD00909.1
CHMPJFD00909.250EA 0 EUR
CHMPJFD00909.250 MAYBJFD00909.1
1-Chlor-2,6-difluor-3-nitrobenzol
1-Chloro-2,6-difluoro-3-nitrobenzene
Formel: C₆H₂ClF₂NO₂
Molecular Weight: 193,54 g/mol
MDL: MFCD00042201
CAS-Nummer: 3847-58-3
EINECS: 411-980-8
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR