2-Chlor-5-nitrobenzonitril

Lieferant: COMBI-BLOCKS

AN-1050-5G AN-1050-100G AN-1050-25G
COBBAN-1050-5GEA 26.2 EUR
COBBAN-1050-5G COBBAN-1050-100G COBBAN-1050-25G
2-Chlor-5-nitrobenzonitril
2-Chlor-5-nitrobenzonitril
Formel: C₇H₃ClN₂O₂
Molecular Weight: 182,57 g/mol
Siedepunkt: 119…122 °C (0,6 mmHg)
Schmelzpunkt: 108 °C
MDL: MFCD00007292
CAS-Nummer: 16588-02-6
EINECS: 240-643-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR