2-Chlor-3,4-dimethoxybenzonitril
Lieferant: Thermo Scientific
Synonyme:
2-Chlor-3,4-dimethoxybenzoesäurenitril
BTB09913.1
BTB09913.10
BTB09913.50
MAYBBTB09913.1EA
0
EUR
MAYBBTB09913.1
MAYBBTB09913.10
MAYBBTB09913.50
2-Chlor-3,4-dimethoxybenzonitril
2-Chlor-3,4-dimethoxybenzonitril
Formel:
C₉H₈ClNO₂ Molecular Weight: 197.62 g/mol |
MDL:
MFCD00173893 CAS-Nummer: 119413-61-5 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...