2-Chlor-4-hydroxybenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

346071-5G 346071-10G
SIAL346071-5GEA 143 EUR
SIAL346071-5G SIAL346071-10G
2-Chlor-4-hydroxybenzonitril, Sigma-Aldrich®
2-Chlor-4-hydroxybenzonitril
Formel: C₇H₄ClNO
Molecular Weight: 153,57 g/mol
MDL: MFCD00052185
CAS-Nummer: 3336-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR