2-Chlor-3-methoxybenzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlor-3-methoxybenzoesäurenitril

M-Clarity™ Qualitätsstufe = MQ200

657913-10G 657913-1G
SIAL657913-10GEA 385 EUR
SIAL657913-10G SIAL657913-1G
2-Chlor-3-methoxybenzonitril, Sigma-Aldrich®
2-Chlor-3-methoxybenzonitril
Formel: C₈H₆ClNO
Molecular Weight: 167,59 g/mol
CAS-Nummer: 853331-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR