2-Chlor-5-(trifluormethyl)phenol
Lieferant: FLUOROCHEM
Synonyme:
4-Chlor-3-hydroxybenzotrifluorid
004474-5G
004474-25G
FLUO004474-5GEA
42.9
EUR
FLUO004474-5G
FLUO004474-25G
2-Chlor-5-(trifluormethyl)phenol
2-Chlor-5-(trifluormethyl)phenol
Formel:
C₇H₄ClF₃O Molecular Weight: 196.56 g/mol Siedepunkt: 87…88 °C (38 mmHg) Dichte: 1.459 g/cm³ (20 °C) |
MDL:
MFCD00019995 CAS-Nummer: 40889-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...