1-Chlor-3,4-dibrom-5-fluorbenzol
Lieferant: FLUOROCHEM
Synonyme:
5-Chlor-2,3-dibrom-1-fluorbenzol, 5-Chlor-1,2-dibrom-3-fluorbenzol
008671-5G
008671-25G
FLUO008671-5GEA
39.9
EUR
FLUO008671-5G
FLUO008671-25G
1-Chlor-3,4-dibrom-5-fluorbenzol
1-Chloro-3,4-dibromo-5-fluorobenzene
Formel:
C₆H₂Br₂ClF Siedepunkt: 242 °C (1013 hPa) Schmelzpunkt: 98 °C |
MDL:
MFCD00143441 CAS-Nummer: 208186-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...