1-Chlor-3,4-dibrom-5-fluorbenzol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Chlor-2,3-dibrom-1-fluorbenzol, 5-Chlor-1,2-dibrom-3-fluorbenzol

A19536.06 A19536.14
A19536.06EA 46 EUR
A19536.06 A19536.14
1-Chlor-3,4-dibrom-5-fluorbenzol ≥98%
1-Chloro-3,4-dibromo-5-fluorobenzene

Purity: 98% CAS No.: 208186-78-1 Molecular Formula: C6H2Br2ClF Formula Weight: 288.35 Melting Point: 37-39° MDL No.: MFCD00143441

Formel: C₆H₂Br₂ClF
Molecular Weight: 288.34 g/mol
Siedepunkt: 242
Schmelzpunkt: 37…39 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143441
CAS-Nummer: 208186-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR