2'-Chlor-4',5'-difluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(2-Chloro-4,5-difluorophenyl)ethanone, 1-chloro-3,4-difluor-6-ethanoylbenzene

021889-5G 021889-25G
FLUO021889-5GEA 39.9 EUR
FLUO021889-5G FLUO021889-25G
2'-Chlor-4',5'-difluoracetophenon
2'-Chlor-4',5'-difluoracetophenon
Formel: C₈H₅ClF₂O
MDL: MFCD00671761
CAS-Nummer: 121872-94-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR