3-Chlor-5-fluorbenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

655171-25G 655171-5G
SIAL655171-25GEA 112 EUR
SIAL655171-25G SIAL655171-5G
3-Chlor-5-fluorbenzonitril, Sigma-Aldrich®
3-Chlor-5-fluorbenzonitril
Formel: C₇H₃ClFN
Molecular Weight: 155,56 g/mol
MDL: MFCD03407967
CAS-Nummer: 327056-73-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR