3-Chlor-4-fluorbenzonitril ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Chlor-3-cyan-6-fluorbenzene

A15987.06 A15987.14
A15987.06EA 52.7 EUR
A15987.06 A15987.14
3-Chlor-4-fluorbenzonitril ≥98%
3-Chlor-4-fluorbenzonitril

Purity: 98+% CAS No.: 117482-84-5 Molecular Formula: C7H3ClFN Formula Weight: 155.56 Melting Point: 68-72° Beilstein Reference: 4740501 UN No.: 3439 MDL No.: MFCD00015431

Formel: C₇H₃ClFN
Molecular Weight: 155,56 g/mol
Siedepunkt: 135 °C (65 mmHg)
Schmelzpunkt: 68…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015431
CAS-Nummer: 117482-84-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR