3-Chlor-4-fluorbenzonitril

Lieferant: Thermo Scientific

Synonyme: 1-Chlor-3-cyan-6-fluorbenzene

BTB03621.1 BTB03621.10 BTB03621.50
MAYBBTB03621.1EA 0 EUR
MAYBBTB03621.1 MAYBBTB03621.10 MAYBBTB03621.50
3-Chlor-4-fluorbenzonitril
3-Chlor-4-fluorbenzonitril
Formel: C₇H₃ClFN
Molecular Weight: 155,56 g/mol
Siedepunkt: 135 °C (65 mmHg)
Schmelzpunkt: 70…71 °C
MDL: MFCD00015431
CAS-Nummer: 117482-84-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR