3-Chlor-4-hydroxybenzonitril 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H50434.03
H50434.06
H50434.03EA
32
EUR
H50434.03
H50434.06
3-Chlor-4-hydroxybenzonitril 95%
3-Chlor-4-hydroxybenzonitril
Purity: 95%
CAS No.: 2315-81-3
Molecular Formula: C7H4ClNO
Formula Weight: 153.57
Melting Point: 147-149°
MDL No.: MFCD01567246
Formel:
C₇H₄ClNO Schmelzpunkt: 147…149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01567246 CAS-Nummer: 2315-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...