5'-Chlor-2'-hydroxy-3'-nitroacetophenon
Lieferant: CHEMPUR
Synonyme:
1-Chlor-3-ethanoyl-4-hydroxy-5-nitrobenzene, 1-(5-Chloro-2-hydroxy-3-nitrophenyl)ethanone
FL12080.POR
012080-25G
CHMPFL12080.POREA
0
EUR
CHMPFL12080.POR
FLUO012080-25G
5'-Chlor-2'-hydroxy-3'-nitroacetophenon
5'-Chlor-2'-hydroxy-3'-nitroacetophenon
Formel:
C₈H₆ClNO₄ Molecular Weight: 215,59 g/mol |
MDL:
MFCD00191921 CAS-Nummer: 84942-40-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...