3'-Chlor-4'-fluoracetophenon
Lieferant: Thermo Scientific
TL00329.10
TL00329.50
MAYBTL00329.10EA
0
EUR
MAYBTL00329.10
MAYBTL00329.50
3'-Chlor-4'-fluoracetophenon
3'-Chlor-4'-fluoracetophenon
Formel:
C₈H₆ClFO Molecular Weight: 172,59 g/mol |
MDL:
MFCD00042203 CAS-Nummer: 2923-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...