3-Chlor-5-fluorphenol
Lieferant: COMBI-BLOCKS
AN-2568-5G
AN-2568-25G
AN-2568-1G
COBBAN-2568-5GEA
193
EUR
COBBAN-2568-5G
COBBAN-2568-25G
COBBAN-2568-1G
3-Chlor-5-fluorphenol
3-Chlor-5-fluorphenol
Formel:
C₆H₄ClFO Molecular Weight: 146,55 g/mol |
MDL:
MFCD00042212 CAS-Nummer: 202982-70-5 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...