3-Chlor-p-kresol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlor-4-hydroxytoluol, 3-Chlor-4-methylphenol, 1-Chlor-3-hydroxy-6-methylbenzol

708224-1G 708224-5G
SIAL708224-1GEA 24.4 EUR
SIAL708224-1G SIAL708224-5G
3-Chlor-p-kresol, Sigma-Aldrich®
3-Chlor-p-kresol
Formel: C₇H₇ClO
Molecular Weight: 142,58 g/mol
MDL: MFCD00060319
CAS-Nummer: 615-62-3
EINECS: 210-439-1
UN: 3437
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR