4-Chlor-3-nitrobenzonitril

Lieferant: COMBI-BLOCKS

AN-1053-1G AN-1053-25G AN-1053-5G
COBBAN-1053-1GEA 0 EUR
COBBAN-1053-1G COBBAN-1053-25G COBBAN-1053-5G
4-Chlor-3-nitrobenzonitril
4-Chlor-3-nitrobenzonitril
Formel: C₇H₃ClN₂O₂
Molecular Weight: 182,57 g/mol
Schmelzpunkt: 100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016987
CAS-Nummer: 939-80-0
EINECS: 213-364-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR