4'-Chlor-3'-(trifluormethyl)acetophenon
Lieferant: FLUOROCHEM
Synonyme:
4-Chloro-3-(trifluoromethyl)acetophenone
013009-5G
013009-1G
FLUO013009-5GEA
104
EUR
FLUO013009-5G
FLUO013009-1G
4'-Chlor-3'-(trifluormethyl)acetophenon
4'-Chlor-3'-(trifluormethyl)acetophenon
Formel:
C₉H₆ClF₃O Molecular Weight: 222.59 g/mol |
MDL:
MFCD03094146 CAS-Nummer: 129825-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...