4'-Chlor-2'-fluoracetophenon

Lieferant: CHEMPUR

FL13255.POR 13255-1G 013255-5G 013255-1G
CHMPFL13255.POREA 0 EUR
CHMPFL13255.POR FLUO13255-1G FLUO013255-5G FLUO013255-1G
4'-Chlor-2'-fluoracetophenon
4'-Chlor-2'-fluoracetophenon
Formel: C₈H₆ClFO
Molecular Weight: 172.59 g/mol
MDL: MFCD04116328
CAS-Nummer: 175711-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR