1-Chlor-4-ethinylbenzol

Lieferant: COMBI-BLOCKS

Synonyme: (4-Chlorphenyl)acetylen, 1-Ethinyl-4-chlorbenzol, (4-Chlorphenyl)ethin

OS-7584-5G OS-7584-1G OS-7584-25G
COBBOS-7584-5GEA 72.8 EUR
COBBOS-7584-5G COBBOS-7584-1G COBBOS-7584-25G
1-Chlor-4-ethinylbenzol
1-Chlor-4-ethinylbenzol
Formel: C₈H₅Cl
Molecular Weight: 136.58 g/mol
MDL: MFCD00191917
CAS-Nummer: 873-73-4
UN: 1325
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR