4-Chlor-3-(trifluormethyl)phenol

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-5-hydroxybenzotrifluorid

004472-5G 004472-25G
FLUO004472-5GEA 34.1 EUR
FLUO004472-5G FLUO004472-25G
4-Chlor-3-(trifluormethyl)phenol
4-Chlor-3-(trifluormethyl)phenol
Formel: C₇H₄ClF₃O
MDL: MFCD00042525
CAS-Nummer: 6294-93-5
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR