2,3-Difluor-4-hydroxybenzonitril
Lieferant: CHEMPUR
CFF1025.0.5
FL13038.POR
013038-1G
013038-5G
CHMPCFF1025.0.5EA
401
EUR
CHMPCFF1025.0.5
CHMPFL13038.POR
FLUO013038-1G
FLUO013038-5G
2,3-Difluor-4-hydroxybenzonitril
2,3-Difluor-4-hydroxybenzonitril
Formel:
C₇H₃F₂NO Molecular Weight: 155.1 g/mol |
MDL:
MFCD00269596 CAS-Nummer: 126162-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...