2,6-Difluor-4-hydroxybenzonitril

Lieferant: CHEMPUR

05-0024.1 05-0024.5
CHMP05-0024.1EA 0 EUR
CHMP05-0024.1 CHMP05-0024.5
2,6-Difluor-4-hydroxybenzonitril
2,6-Difluor-4-hydroxybenzonitril
Formel: C₇H₃F₂NO
MDL: MFCD03094499
CAS-Nummer: 123843-57-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR