2,6-Difluor-4-hydroxybenzonitril

Lieferant: CHEMPUR

Synonyme: 4-Cyano-3,5-difluorophenol

FL13816.POR 013816-1G 013816-5G
CHMPFL13816.POREA 0 EUR
CHMPFL13816.POR FLUO013816-1G FLUO013816-5G
2,6-Difluor-4-hydroxybenzonitril
2,6-Difluor-4-hydroxybenzonitril
Formel: C₇H₃F₂NO
Molecular Weight: 155.1 g/mol
MDL: MFCD03094499
CAS-Nummer: 123843-57-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR