2,6-Difluor-4-methoxybenzonitril

Lieferant: CHEMPUR

Synonyme: 2,6-Difluor-4-methoxybenzoesäurenitril

05-0017.1 05-0017.5
CHMP05-0017.1EA 0 EUR
CHMP05-0017.1 CHMP05-0017.5
2,6-Difluor-4-methoxybenzonitril
2,6-Difluor-4-methoxybenzonitril
Formel: C₈H₅F₂NO
Molecular Weight: 169,13 g/mol
MDL: MFCD03094498
CAS-Nummer: 123843-66-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR