2-Fluor-4-methoxybenzonitril

Lieferant: CHEMPUR

FL13330.POR 013330-1G 013330-5G
CHMPFL13330.POREA 0 EUR
CHMPFL13330.POR FLUO013330-1G FLUO013330-5G
2-Fluor-4-methoxybenzonitril
2-Fluor-4-methoxybenzonitril
Formel: C₈H₆FNO
MDL: MFCD00509374
CAS-Nummer: 94610-82-9
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR