2-Fluor-5-methoxybenzonitril

Lieferant: CHEMPUR

Synonyme: 2-Fluor-5-methoxybenzoesäurenitril

FL13331.POR 013331-5G 013331-1G 013331-25G
CHMPFL13331.POREA 0 EUR
CHMPFL13331.POR FLUO013331-5G FLUO013331-1G FLUO013331-25G
2-Fluor-5-methoxybenzonitril
2-Fluor-5-methoxybenzonitril
Formel: C₈H₆FNO
Molecular Weight: 151,14 g/mol
MDL: MFCD00142921
CAS-Nummer: 127667-01-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR